Gene Transcripts

Gene Symbol
Olr1443
Entrez Gene ID
287331
Gene Name
olfactory receptor 1443
Gene Aliases
Chromosome Location
Chr. 10: 44547564 - 44548484 on Build Rnor_6.0
UniGene ID
Rn.142132
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000018.1 NP_001000018.1 1 834

View Full Product Details