Gene Transcripts

Gene Symbol
U2surp
Entrez Gene ID
315903
Gene Name
U2 snRNP-associated SURP domain containing
Gene Aliases
RGD1307882
Chromosome Location
Chr. 8: 103242095 - 103299070 on Build Rnor_6.0
UniGene ID
Rn.208353
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002729954.4 XP_002730000.1 25 2714
XM_006243554.1 XP_006243616.1 26 2859
XM_236501.7 XP_236501.3 26 2900

View Full Product Details