Gene Transcripts

Gene Symbol
Olr1400
Entrez Gene ID
405991
Gene Name
olfactory receptor 1400
Gene Aliases
Chromosome Location
Chr. 10: 34771715 - 34772570 on Build Rnor_6.0
UniGene ID
Rn.142167
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001096.1 NP_001001096.1 672

Gene Symbol
Olr1399
Entrez Gene ID
405990
Gene Name
olfactory receptor 1399
Gene Aliases
Chromosome Location
Chr. 10: 34756094 - 34757029 on Build Rnor_6.0
UniGene ID
Rn.142168
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001095.1 NP_001001095.1 1 672

Gene Symbol
Olr1401
Entrez Gene ID
287247
Gene Name
olfactory receptor 1401
Gene Aliases
ratchr10-34999096-35000031_ORF
Chromosome Location
Chr. 10: 34804878 - 34805813 on Build Rnor_6.0
UniGene ID
Rn.142166
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000003.1 NP_001000003.1 1 672

View Full Product Details