Gene Transcripts
- Gene Symbol
- Rpl17
- Entrez Gene ID
- 319195
- Gene Name
- ribosomal protein L17
- Gene Aliases
- Chromosome Location
- Chr. 18: 74998978 - 75003381 on Build GRCm38
- UniGene ID
- Mm.276337
- Species
- Mus musculus
- Gene Symbol
- Hk1
- Entrez Gene ID
- 15275
- Gene Name
- hexokinase 1
- Gene Aliases
- BB404130, Hk-1, Hk1-s, dea, mHk1-s
- Chromosome Location
- Chr. 10: 62268855 - 62379920 on Build GRCm38
- UniGene ID
- Mm.196605
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
J05277.1 |
- |
|
3700 |
|
View Full Product Details