Gene Transcripts

Gene Symbol
Olfr1412
Entrez Gene ID
258274
Gene Name
olfactory receptor 1412
Gene Aliases
MOR208-4
Chromosome Location
Chr. 1: 92588332 - 92589297 on Build GRCm38
UniGene ID
Mm.222974
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146277.1 NP_666389.1 1 607
GenBank mRNA BC125241.1 - 1 607

View Full Product Details