Gene Transcripts

Gene Symbol
LOC679038
Entrez Gene ID
679038
Gene Name
similar to mammalian retrotransposon derived 8b
Gene Aliases
Chromosome Location
Chr. X: 73616371 - 73617584 on Build Rnor_6.0
UniGene ID
Rn.202610
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100998.1 NP_001094468.1 1 1069
GenBank mRNA BC105915.1 - 1 1069

View Full Product Details