Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001083958.1 | NP_001077427.1 | 2 | 239 | |
NM_028298.3 | NP_082574.1 | 2 | 195 | ||
GenBank mRNA | AK012336.1 | - | 2 | 198 | |
AK018500.1 | - | 2 | 230 | ||
AK161949.1 | - | 2 | 241 | ||
AK220552.1 | - | 2 | 200 | ||
BC019153.1 | - | 2 | 205 | ||
BC034731.1 | - | 2 | 198 | ||
BC060660.1 | - | 2 | 231 | 30355779 | |
BC061032.1 | - | 2 | 208 | ||
BC062945.1 | - | 2 | 195 | 30357186 |