Gene Transcripts

Gene Symbol
Gm11567
Entrez Gene ID
670533
Gene Name
predicted gene 11567
Gene Aliases
OTTMUSG00000002206
Chromosome Location
Chr. 11: 99879187 - 99880229 on Build GRCm38
UniGene ID
Mm.459090
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101613.1 NP_001095083.1 1 893
GenBank mRNA AK003994.1 - 1 894

View Full Product Details