Gene Transcripts
- Gene Symbol
- Gp1bb
- Entrez Gene ID
- 116727
- Gene Name
- glycoprotein Ib (platelet), beta polypeptide
- Gene Aliases
- Chromosome Location
- Chr. 11: 86520992 - 86522169 on Build Rnor_6.0
- UniGene ID
- Rn.96497
- Species
- Rattus norvegicus
- Gene Symbol
- Sept5
- Entrez Gene ID
- 116728
- Gene Name
- septin 5
- Gene Aliases
- 5-Sep, CDCrel-1A, Pnutl1
- Chromosome Location
- Chr. 11: 86516390 - 86522169 on Build Rnor_6.0
- UniGene ID
- Rn.96497
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053931.3 |
NP_446383.3
|
11 |
3572 |
|
GenBank mRNA |
AB027143.1 |
- |
11 |
3573 |
|
View Full Product Details