Gene Transcripts

Gene Symbol
SETBP1
Entrez Gene ID
26040
Gene Name
SET binding protein 1
Gene Aliases
MRD29, SEB
Chromosome Location
Chr. 18: 44680173 - 45068510 on Build GRCh38
UniGene ID
Hs.435458
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015559.2 NP_056374.2 4 3311
GenBank mRNA AB007897.2 - 4 3311
AB022660.1 - 3 2858
BC146776.1 - 4 3311 9007225

View Full Product Details