Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022075.4 | NP_071358.1 | 7 | 981 | |
NM_181746.3 | NP_859530.1 | 7 | 1021 | ||
XM_011509451.2 | XP_011507753.1 | 7 | 684 | ||
XM_011509452.2 | XP_011507754.1 | 7 | 732 | ||
GenBank mRNA | AF177338.2 | - | 7 | 710 | |
AF189062.4 | - | 2 | 194 | ||
AK001105.1 | - | 6 | 650 | ||
AK290989.1 | - | 7 | 657 | ||
AY091458.1 | - | 7 | 803 | ||
BC001357.2 | - | 7 | 676 | 3051086 | |
BC010032.2 | - | 7 | 640 | 3506408 | |
BQ219926.1 | - | 7 | 718 | ||
BU543940.1 | - | 7 | 772 |