Gene Transcripts
- Gene Symbol
- ZNF415
- Entrez Gene ID
- 55786
- Gene Name
- zinc finger protein 415
- Gene Aliases
- ZfLp
- Chromosome Location
- Chr. 19: 53107879 - 53133077 on Build GRCh38
- UniGene ID
- Hs.147765
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001136038.2 |
NP_001129510.2
|
3, 4 |
503 |
|
NM_001164309.1 |
NP_001157781.1
|
3, 4 |
376 |
|
NM_018355.3 |
NP_060825.2
|
3, 4 |
271 |
|
XM_006723268.1 |
XP_006723331.1
|
3, 4 |
337 |
|
XM_011527103.1 |
XP_011525405.1
|
4, 5 |
408 |
|
XM_017026964.1 |
XP_016882453.1
|
4, 5 |
491 |
|
XM_017026965.1 |
XP_016882454.1
|
3, 4 |
505 |
|
XM_017026966.1 |
XP_016882455.1
|
4, 5 |
633 |
|
XM_017026967.1 |
XP_016882456.1
|
3, 4 |
431 |
|
GenBank mRNA |
AK002053.1 |
- |
3, 4 |
243 |
|
AY283600.1 |
- |
3, 4 |
243 |
|
DA075573.1 |
- |
3, 4 |
358 |
|
DA518713.1 |
- |
3, 4 |
473 |
|
DB285275.1 |
- |
3, 4 |
377 |
|
View Full Product Details