Gene Transcripts

Gene Symbol
ZACN
Entrez Gene ID
353174
Gene Name
zinc activated ion channel
Gene Aliases
L2, LGICZ, LGICZ1, ZAC, ZAC1
Chromosome Location
Chr. 17: 76079182 - 76082804 on Build GRCh38
UniGene ID
Hs.714919
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_180990.3 NP_851321.2 5, 6 627
GenBank mRNA AB223030.1 - 5, 6 541
AF512521.1 - 5, 6 581
BC110597.1 - 5, 6 581 40023955

View Full Product Details