Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042659.1 | NP_001036124.1 | 2 | 1545 | |
NM_022721.3 | NP_073558.2 | 2 | 1652 | ||
GenBank mRNA | AB052910.1 | - | 1 | 778 | |
AF005203.1 | - | 1 | 152 | ||
AF272146.1 | - | 1 | 769 | ||
AK030111.1 | - | 2 | 1654 | ||
AK039402.1 | - | 2 | 1545 | ||
BC117723.2 | - | 2 | 1078 | 40088671 |