Gene Transcripts
- Gene Symbol
- Uqcrq
- Entrez Gene ID
- 22272
- Gene Name
- ubiquinol-cytochrome c reductase, complex III subunit VII
- Gene Aliases
- 1100001F06Rik, 1500040F11Rik, 5830407L17Rik, 9.5kDa, AA959903, QP-C, Qpc, Uqcrb, c1502
- Chromosome Location
- Chr. 11: 53428948 - 53430874 on Build GRCm38
- UniGene ID
- Mm.251621
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK005374.1 |
- |
|
826 |
|
View Full Product Details