Gene Transcripts

Gene Symbol
Dgke
Entrez Gene ID
56077
Gene Name
diacylglycerol kinase, epsilon
Gene Aliases
C87606, DAGK6, DGK
Chromosome Location
Chr. 11: 89037582 - 89060763 on Build GRCm38
UniGene ID
Mm.153695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019505.3 NP_062378.1 7 1508
GenBank mRNA AK049335.1 - 7 1308
AK147841.1 - 7 1509

View Full Product Details