Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029789.1 | NP_084065.1 | 6 | 586 | |
GenBank mRNA | AK002626.1 | - | 6 | 553 | |
AK050161.1 | - | 6 | 567 | ||
AK146106.1 | - | 6 | 565 | ||
AK159325.1 | - | 6 | 565 | ||
AY029532.1 | - | 6 | 586 | ||
BC006847.1 | - | 6 | 558 | 3598473 |