Gene Transcripts

Gene Symbol
Ern1
Entrez Gene ID
78943
Gene Name
endoplasmic reticulum (ER) to nucleus signalling 1
Gene Aliases
9030414B18Rik, AI225830, C85377, Ire1a, Ire1alpha, Ire1p
Chromosome Location
Chr. 11: 106397620 - 106487796 on Build GRCm38
UniGene ID
Mm.340943
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023913.2 NP_076402.1 19 2541
GenBank mRNA AB031332.1 - 19 2541
AK077041.1 - 2564
AK080709.1 - 13 1798

View Full Product Details