Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317335.1 | NP_001304264.1 | 5 | 855 | |
NM_001928.3 | NP_001919.2 | 5 | 834 | ||
GenBank mRNA | AK300963.1 | - | 5 | 829 | |
BC034529.1 | - | 4 | 750 | ||
BC040146.1 | - | 4 | 736 | ||
BC051001.1 | - | 5 | 774 | ||
BI763909.1 | - | 210 | |||
M84526.1 | - | 5 | 782 |