Gene Transcripts

Gene Symbol
PPIL2
Entrez Gene ID
23759
Gene Name
peptidylprolyl isomerase like 2
Gene Aliases
CYC4, CYP60, Cyp-60, UBOX7, hCyP-60
Chromosome Location
Chr. 22: 21665984 - 21698276 on Build GRCh38
UniGene ID
Hs.438587
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317996.1 NP_001304925.1 10 697
NM_014337.3 NP_055152.1 10 697
NM_148175.2 NP_680480.1 10 697
NM_148176.2 NP_680481.1 10 697
XM_005261448.3 XP_005261505.1 10 697
XM_011530041.2 XP_011528343.1 10 697
XM_011530042.2 XP_011528344.1 10 697
XM_011530043.2 XP_011528345.1 10 697
XM_011530044.2 XP_011528346.1 10 697
XM_011530046.2 XP_011528348.1 10 697
XM_011530047.2 XP_011528349.1 10 697
XM_011530049.2 XP_011528351.1 11 747
XM_011530050.2 XP_011528352.1 9 610
XM_011530051.2 XP_011528353.1 10 637
GenBank mRNA AK127259.1 - 11 720
AK289545.1 - 10 659
AK299206.1 - 9 559
BC000022.2 - 10 659 3505936
BC028385.2 - 10 641 4816793
CR456548.1 - 10 613
U37219.1 - 10 673

View Full Product Details