Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002451.3 | NP_002442.2 | 4 | 367 | |
GenBank mRNA | AF109294.1 | - | 4 | 375 | |
AK300592.1 | - | 4 | 530 | ||
AK309365.1 | - | 4 | 354 | ||
AY712791.1 | - | 4 | 375 | ||
BC012316.1 | - | 4 | 341 | 3546198 | |
BC018625.1 | - | 331 | 3030148 | ||
BC026106.1 | - | 4 | 349 | 4820938 | |
CR541670.1 | - | 4 | 254 | ||
CR541710.1 | - | 4 | 254 | ||
FJ224338.1 | - | 4 | 254 | ||
HE654772.1 | - | 4 | 363 | ||
HE654773.1 | - | 4 | 363 | ||
HE654774.1 | - | 4 | 363 | ||
HE654775.1 | - | 4 | 363 | ||
HE654776.1 | - | 4 | 363 | ||
HE654777.1 | - | 4 | 363 | ||
L40432.1 | - | 4 | 364 | ||
U22233.1 | - | 4 | 375 |