Gene Transcripts

Gene Symbol
EPB41
Entrez Gene ID
2035
Gene Name
erythrocyte membrane protein band 4.1
Gene Aliases
4.1R, EL1, HE
Chromosome Location
Chr. 1: 28887091 - 29120046 on Build GRCh38
UniGene ID
Hs.175437
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166005.1 NP_001159477.1 7 1218
NM_001166006.1 NP_001159478.1 7 1291
NM_001166007.1 NP_001159479.1 7 1201
NM_004437.3 NP_004428.1 8 1281
NM_203342.2 NP_976217.1 8 1298
NM_203343.2 NP_976218.1 6 1113
XM_005245753.1 XP_005245810.1 7 1293
XM_005245757.1 XP_005245814.1 7 1293
XM_005245760.1 XP_005245817.1 7 1293
XM_005245761.1 XP_005245818.1 7 1293
XM_005245763.1 XP_005245820.1 7 1293
XM_005245764.1 XP_005245821.1 7 1293
XM_005245765.1 XP_005245822.1 7 1293
XM_005245768.1 XP_005245825.1 7 1293
XM_005245769.1 XP_005245826.1 7 1293
XM_005245770.1 XP_005245827.1 7 1293
XM_005245772.4 XP_005245829.1 7 1293
XM_005245773.4 XP_005245830.1 7 1293
XM_005245774.1 XP_005245831.1 7 1293
XM_006710434.1 XP_006710497.1 7 1293
XM_006710439.1 XP_006710502.1 7 1293
XM_011540956.1 XP_011539258.1 7 1293
XM_011540957.1 XP_011539259.1 7 1293
XM_011540958.1 XP_011539260.1 7 1293
XM_011540959.1 XP_011539261.1 7 1293
XM_011540960.1 XP_011539262.1 7 1293
XM_011540961.1 XP_011539263.1 7 1293
XM_011540962.1 XP_011539264.1 7 1293
XM_011540963.2 XP_011539265.1 7 1293
XM_011540964.1 XP_011539266.1 7 1189
XM_011540965.2 XP_011539267.1 7 1293
XM_017000581.1 XP_016856070.1 6 1188
XM_017000582.1 XP_016856071.1 6 1188
XM_017000583.1 XP_016856072.1 7 1293
XM_017000584.1 XP_016856073.1 7 1584
XM_017000585.1 XP_016856074.1 7 1581
XM_017000586.1 XP_016856075.1 7 1293
XM_017000587.1 XP_016856076.1 7 1293
XM_017000588.1 XP_016856077.1 7 1293
XM_017000589.1 XP_016856078.1 6 1188
XM_017000590.1 XP_016856079.1 6 1188
XM_017000591.1 XP_016856080.1 7 1293
XM_017000592.1 XP_016856081.1 7 1293
XM_017000593.1 XP_016856082.1 7 1189
XM_017000594.1 XP_016856083.1 7 1189
XM_017000595.1 XP_016856084.1 7 1189
XM_017000596.1 XP_016856085.1 7 1189
XM_017000597.1 XP_016856086.1 7 1189
XM_017000598.1 XP_016856087.1 7 1189
XM_017000599.1 XP_016856088.1 7 1189
XM_017000600.1 XP_016856089.1 7 1189
XM_017000601.1 XP_016856090.1 7 1189
XM_017000602.1 XP_016856091.1 7 1189
XM_017000603.1 XP_016856092.1 7 1189
XM_017000604.1 XP_016856093.1 7 1189
GenBank mRNA AB209649.1 - 7 1201
AF156225.1 - 6 1091
AK310655.1 - 6 1129
BC039079.1 - 7 1288 5167140
BC096104.1 - 4 507
BC096105.1 - 6 825 40001729
BC113841.1 - 6 825
BC114450.1 - 6 825
J03796.1 - 6 1032
M14993.1 - 8 1262
M61733.1 - 8 1279

View Full Product Details