Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007805.4 | NP_031831.2 | 6 | 2259 | |
GenBank mRNA | AK014395.2 | - | 6 | 2260 | |
AK028780.1 | - | 5 | 2133 | ||
AK158978.1 | - | 7 | 2590 | ||
AK169711.1 | - | 6 | 2248 | ||
BC006732.1 | - | 6 | 2299 | 3963612 |