Gene Transcripts
- Gene Symbol
- Arhgef11
- Entrez Gene ID
- 78966
- Gene Name
- Rho guanine nucleotide exchange factor (GEF) 11
- Gene Aliases
- Gtrap48
- Chromosome Location
- Chr. 2: 186979850 - 187102523 on Build Rnor_6.0
- UniGene ID
- Rn.229365
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023982.1 |
NP_076472.1
|
9, 10 |
629 |
|
GenBank mRNA |
AF225961.1 |
- |
9, 10 |
629 |
|
View Full Product Details