Gene Transcripts

Gene Symbol
NUDT16
Entrez Gene ID
131870
Gene Name
nudix hydrolase 16
Gene Aliases
Chromosome Location
Chr. 3: 131381671 - 131388830 on Build GRCh38
UniGene ID
Hs.282050
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171905.1 NP_001165376.1 4 1323
NM_152395.2 NP_689608.2 3 1497
NR_033268.1 2 1584
GenBank mRNA AK055827.1 - 3 1399

View Full Product Details