Gene Transcripts
- Gene Symbol
- NAXE
- Entrez Gene ID
- 128240
- Gene Name
- NAD(P)HX epimerase
- Gene Aliases
- AIBP, APOA1BP, YJEFN1
- Chromosome Location
- Chr. 1: 156591762 - 156599818 on Build GRCh38
- UniGene ID
- Hs.528320
- Species
- Homo sapiens
- Gene Symbol
- GPATCH4
- Entrez Gene ID
- 54865
- Gene Name
- G-patch domain containing 4
- Gene Aliases
- GPATC4
- Chromosome Location
- Chr. 1: 156594308 - 156601496 on Build GRCh38
- UniGene ID
- Hs.193832
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055377.1 |
- |
1 |
2139 |
|
View Full Product Details