Gene Transcripts
- Gene Symbol
- YEATS2
- Entrez Gene ID
- 55689
- Gene Name
- YEATS domain containing 2
- Gene Aliases
- Chromosome Location
- Chr. 3: 183697797 - 183812625 on Build GRCh38
- UniGene ID
- Hs.632575
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK090720.1 |
- |
1 |
756 |
|
View Full Product Details