Gene Transcripts

Gene Symbol
KCNMA1
Entrez Gene ID
3778
Gene Name
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
Chromosome Location
Chr. 10: 76869601 - 77637819 on Build GRCh38
UniGene ID
Hs.144795
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014797.2 NP_001014797.1 29 3782
NM_001271518.1 NP_001258447.1 28 3620
NM_001322829.1 NP_001309758.1 29 3776
NM_001322830.1 NP_001309759.1 31 3872
NM_001322832.1 NP_001309761.1 28 3767
NM_001322835.1 NP_001309764.1 30 3860
NM_001322838.1 NP_001309767.1 30 3814
XM_005269776.3 XP_005269833.1 30 3866
XM_005269778.1 XP_005269835.1 30 3942
XM_005269781.2 XP_005269838.1 29 3865
XM_005269787.3 XP_005269844.1 29 3785
XM_005269789.2 XP_005269846.1 30 3784
XM_005269792.2 XP_005269849.1 29 3778
XM_005269796.2 XP_005269853.1 28 3691
XM_011539773.1 XP_011538075.1 31 4077
XM_011539775.1 XP_011538077.1 30 4065
XM_011539776.1 XP_011538078.1 30 4062
XM_011539780.1 XP_011538082.1 30 3945
XM_011539781.2 XP_011538083.1 30 3903
XM_011539785.1 XP_011538087.1 30 3846
XM_017016207.1 XP_016871696.1 32 4158
XM_017016208.1 XP_016871697.1 32 4155
XM_017016210.1 XP_016871699.1 31 4074
XM_017016211.1 XP_016871700.1 31 3987
XM_017016212.1 XP_016871701.1 31 3984
XM_017016213.1 XP_016871702.1 30 3975
XM_017016216.1 XP_016871705.1 30 3900
XM_017016217.1 XP_016871706.1 29 3840
XM_017016223.1 XP_016871712.1 27 3152
GenBank mRNA AB209488.1 - 28 3189
AK310379.1 - 30 3814
BX648925.1 - 25 2932

View Full Product Details