Gene Transcripts
- Gene Symbol
- RPL13
- Entrez Gene ID
- 6137
- Gene Name
- ribosomal protein L13
- Gene Aliases
- BBC1, D16S444E, D16S44E, L13
- Chromosome Location
- Chr. 16: 89560657 - 89566829 on Build GRCh38
- UniGene ID
- Hs.410817
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000977.3 |
NP_000968.2
|
4, 5 |
496 |
|
NM_001243130.1 |
NP_001230059.1
|
4, 5 |
439 |
|
NM_001243131.1 |
NP_001230060.1
|
5, 6 |
355 |
|
NM_033251.2 |
NP_150254.1
|
3, 4 |
669 |
|
GenBank mRNA |
AB062392.1 |
- |
3, 4 |
445 |
|
AK025405.1 |
- |
1 |
490 |
|
AK127579.1 |
- |
4, 5 |
473 |
|
AK130963.1 |
- |
4, 5 |
392 |
|
AK130964.1 |
- |
4, 5 |
341 |
|
AK290893.1 |
- |
4, 5 |
471 |
|
AK294655.1 |
- |
4, 5 |
352 |
|
AK297198.1 |
- |
5, 6 |
331 |
|
AK308545.1 |
- |
3, 4 |
311 |
|
BC000851.1 |
- |
1 |
449 |
|
BC004954.1 |
- |
1 |
459 |
3623320 |
BC006179.1 |
- |
4, 5 |
459 |
|
BC007345.2 |
- |
4, 5 |
461 |
3688320 |
BC007563.1 |
- |
3, 4 |
657 |
2989021 |
BC007805.2 |
- |
4, 5 |
457 |
4298327 |
BC010994.2 |
- |
3, 4 |
452 |
3842093 |
BC013078.1 |
- |
4, 5 |
457 |
3459415 |
BC014167.2 |
- |
1 |
457 |
4550361 |
BC020804.1 |
- |
3, 4 |
669 |
4276103 |
BC027463.1 |
- |
4, 5 |
457 |
5111302 |
BC063378.1 |
- |
1 |
460 |
3542514 |
BC066320.1 |
- |
4, 5 |
471 |
5300601 |
BC093063.1 |
- |
1 |
473 |
30401947 |
BC106058.1 |
- |
4, 5 |
496 |
4992467 |
BG706207.1 |
- |
4, 5 |
418 |
|
X64707.1 |
- |
4, 5 |
471 |
|
- Gene Symbol
- RPL13P5
- Entrez Gene ID
- 283345
- Gene Name
- ribosomal protein L13 pseudogene 5
- Gene Aliases
- RPL13-2, RPL13L, RPL13_4_1199, RRPL13L
- Chromosome Location
- Chr. 12: 6883981 - 6884614 on Build GRCh38
- UniGene ID
- Hs.720698
- Species
- Homo sapiens
View Full Product Details