Gene Transcripts

Gene Symbol
SPIDR
Entrez Gene ID
23514
Gene Name
scaffolding protein involved in DNA repair
Gene Aliases
KIAA0146
Chromosome Location
Chr. 8: 47260878 - 47736001 on Build GRCh38
UniGene ID
Hs.381058
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080394.3 NP_001073863.1 4, 5 452
NM_001282916.1 NP_001269845.1 3, 4 385
NM_001282919.1 NP_001269848.1 4, 5 415
NR_104581.1 4, 5 452
XM_005251189.3 XP_005251246.1 4, 5 394
XM_005251191.3 XP_005251248.1 4, 5 394
XM_005251193.3 XP_005251250.1 4, 5 394
XM_005251198.4 XP_005251255.1 4, 5 423
XM_005251199.4 XP_005251256.1 3, 4 338
XM_006716443.3 XP_006716506.1 4, 5 362
XM_006716444.3 XP_006716507.1 4, 5 386
XM_011517497.2 XP_011515799.1 4, 5 394
XM_017013268.1 XP_016868757.1 4, 5 394
XM_017013269.1 XP_016868758.1 4, 5 394
XM_017013271.1 XP_016868760.1 4, 5 509
XM_017013273.1 XP_016868762.1 4, 5 423
XM_017013277.1 XP_016868766.1 4, 5 379
XM_017013279.1 XP_016868768.1 3, 4 338
XM_017013280.1 XP_016868769.1 4, 5 423
XM_017013281.1 XP_016868770.1 4, 5 388
XM_017013283.1 XP_016868772.1 3, 4 338
XM_017013285.1 XP_016868774.1 4, 5 423
XM_017013286.1 XP_016868775.1 3, 4 338
XM_017013289.1 XP_016868778.1 1, 2 112
XM_017013290.1 XP_016868779.1 3, 4 338
XM_017013291.1 XP_016868780.1 3, 4 338
GenBank mRNA AK122820.1 - 5, 6 750
AK293809.1 - 4, 5 375
AK293972.1 - 4, 5 375
AK294275.1 - 3, 4 307
AK297681.1 - 4, 5 374
AK303579.1 - 4, 5 362
BC031627.1 - 4, 5 377
D63480.1 - 4, 5 380
DR000775.1 - 4, 5 441
HY012281.1 - 4, 5 453

View Full Product Details