Gene Transcripts
- Gene Symbol
- SP140L
- Entrez Gene ID
- 93349
- Gene Name
- SP140 nuclear body protein like
- Gene Aliases
- Chromosome Location
- Chr. 2: 230327152 - 230407506 on Build GRCh38
- UniGene ID
- Hs.589661
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK023116.1 |
- |
10 |
1672 |
|
View Full Product Details