Gene Transcripts

Gene Symbol
PGAM1
Entrez Gene ID
5223
Gene Name
phosphoglycerate mutase 1
Gene Aliases
HEL-S-35, PGAM-B, PGAMA
Chromosome Location
Chr. 10: 97426125 - 97433444 on Build GRCh38
UniGene ID
Hs.632918
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317079.1 NP_001304008.1 4 779
NM_002629.3 NP_002620.1 4 849
GenBank mRNA AK223096.1 - 4 695
AK292216.1 - 4 750
AK293023.1 - 4 694
AK295991.1 - 5 585
AK296619.1 - 4 638
AK297491.1 - 5 595
AK312351.1 - 4 678
AK315910.1 - 4 782
AY007118.1 - 1 686
BC000455.1 - 1 712 2821567
BC010038.1 - 1 704 3604026
BC011678.2 - 1 668 4299008
BC053356.1 - 1 685 5787678
BC062302.1 - 1 684 6584466
BC066959.1 - 4 717 5262935
BC073742.1 - 1 683 6297173
BC157873.1 - 4 701 8862699
CR541659.1 - 4 666
CR541744.1 - 4 666
EU794644.1 - 4 678
J04173.1 - 4 697

Gene Symbol
PGAM4
Entrez Gene ID
441531
Gene Name
phosphoglycerate mutase family member 4
Gene Aliases
PGAM-B, PGAM1, PGAM3, dJ1000K24.1
Chromosome Location
Chr. X: 77967961 - 77969638 on Build GRCh38
UniGene ID
Hs.632822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029891.2 NP_001025062.1 1 666
GenBank mRNA BG875904.1 - 1 518

View Full Product Details