Gene Transcripts
- Gene Symbol
- PGAM1
- Entrez Gene ID
- 5223
- Gene Name
- phosphoglycerate mutase 1
- Gene Aliases
- HEL-S-35, PGAM-B, PGAMA
- Chromosome Location
- Chr. 10: 97426125 - 97433444 on Build GRCh38
- UniGene ID
- Hs.632918
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317079.1 |
NP_001304008.1
|
2, 3 |
520 |
|
NM_002629.3 |
NP_002620.1
|
2, 3 |
590 |
|
GenBank mRNA |
AK223096.1 |
- |
2, 3 |
436 |
|
AK292216.1 |
- |
2, 3 |
491 |
|
AK293023.1 |
- |
2, 3 |
435 |
|
AK295991.1 |
- |
3, 4 |
326 |
|
AK296619.1 |
- |
2, 3 |
379 |
|
AK297491.1 |
- |
3, 4 |
336 |
|
AK312351.1 |
- |
2, 3 |
419 |
|
AK315910.1 |
- |
2, 3 |
523 |
|
AY007118.1 |
- |
1 |
427 |
|
BC000455.1 |
- |
1 |
453 |
2821567 |
BC010038.1 |
- |
1 |
445 |
3604026 |
BC011678.2 |
- |
1 |
409 |
4299008 |
BC053356.1 |
- |
1 |
426 |
5787678 |
BC062302.1 |
- |
1 |
425 |
6584466 |
BC066959.1 |
- |
2, 3 |
458 |
5262935 |
BC073742.1 |
- |
1 |
424 |
6297173 |
BC157873.1 |
- |
2, 3 |
442 |
8862699 |
CR541659.1 |
- |
2, 3 |
407 |
|
CR541744.1 |
- |
2, 3 |
407 |
|
EU794644.1 |
- |
2, 3 |
419 |
|
J04173.1 |
- |
2, 3 |
438 |
|
- Gene Symbol
- PGAM1P5
- Entrez Gene ID
- 100132594
- Gene Name
- phosphoglycerate mutase 1 pseudogene 5
- Gene Aliases
- Chromosome Location
- Chr. 12: 95649255 - 95673999 on Build GRCh38
- UniGene ID
- Hs.676211
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_077225.1 |
|
5 |
859 |
|
GenBank mRNA |
BC022928.1 |
- |
5 |
860 |
|
BX509596.1 |
- |
1 |
148 |
|
- Gene Symbol
- PGAM4
- Entrez Gene ID
- 441531
- Gene Name
- phosphoglycerate mutase family member 4
- Gene Aliases
- PGAM-B, PGAM1, PGAM3, dJ1000K24.1
- Chromosome Location
- Chr. X: 77967961 - 77969638 on Build GRCh38
- UniGene ID
- Hs.632822
- Species
- Homo sapiens
View Full Product Details