Gene Transcripts
- Gene Symbol
- TRIM5
- Entrez Gene ID
- 85363
- Gene Name
- tripartite motif containing 5
- Gene Aliases
- RNF88, TRIM5alpha
- Chromosome Location
- Chr. 11: 5590906 - 5685109 on Build GRCh38
- UniGene ID
- Hs.125300
- Species
- Homo sapiens
- Gene Symbol
- APOE
- Entrez Gene ID
- 348
- Gene Name
- apolipoprotein E
- Gene Aliases
- AD2, APO-E, LDLCQ5, LPG
- Chromosome Location
- Chr. 19: 44905749 - 44909395 on Build GRCh38
- UniGene ID
- Hs.654439
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK308546.1 |
- |
2 |
511 |
|
- Gene Symbol
- ARNTL2
- Entrez Gene ID
- 56938
- Gene Name
- aryl hydrocarbon receptor nuclear translocator like 2
- Gene Aliases
- BMAL2, CLIF, MOP9, PASD9, bHLHe6
- Chromosome Location
- Chr. 12: 27332854 - 27425813 on Build GRCh38
- UniGene ID
- Hs.445447
- Species
- Homo sapiens
- Gene Symbol
- ATP5G2
- Entrez Gene ID
- 517
- Gene Name
- ATP synthase, H+ transporting, mitochondrial Fo complex subunit C2 (subunit 9)
- Gene Aliases
- ATP5A
- Chromosome Location
- Chr. 12: 53665160 - 53677546 on Build GRCh38
- UniGene ID
- Hs.524464
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK075351.1 |
- |
1 |
364 |
|
- Gene Symbol
- CLRN1-AS1
- Entrez Gene ID
- 116933
- Gene Name
- CLRN1 antisense RNA 1
- Gene Aliases
- CLRN1OS, UCRP
- Chromosome Location
- Chr. 3: 150972678 - 151079830 on Build GRCh38
- UniGene ID
- Hs.570680
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126442.1 |
- |
5 |
1569 |
|
- Gene Symbol
- DNAJA3
- Entrez Gene ID
- 9093
- Gene Name
- DnaJ heat shock protein family (Hsp40) member A3
- Gene Aliases
- HCA57, TID1, hTID-1
- Chromosome Location
- Chr. 16: 4425805 - 4456775 on Build GRCh38
- UniGene ID
- Hs.459779
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK127660.1 |
- |
1 |
420 |
|
- Gene Symbol
- FRMD1
- Entrez Gene ID
- 79981
- Gene Name
- FERM domain containing 1
- Gene Aliases
- bA164L23.1
- Chromosome Location
- Chr. 6: 168055745 - 168093007 on Build GRCh38
- UniGene ID
- Hs.266746
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK074110.1 |
- |
5 |
885 |
|
- Gene Symbol
- HAUS3
- Entrez Gene ID
- 79441
- Gene Name
- HAUS augmin like complex subunit 3
- Gene Aliases
- C4orf15, IT1, dgt3
- Chromosome Location
- Chr. 4: 2231836 - 2242164 on Build GRCh38
- UniGene ID
- Hs.665869
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CR749640.1 |
- |
5 |
3840 |
|
- Gene Symbol
- HERC2P4
- Entrez Gene ID
- 100289574
- Gene Name
- hect domain and RLD 2 pseudogene 4
- Gene Aliases
- D16F37S5
- Chromosome Location
- Chr. 16: 32170043 - 32188113 on Build GRCh38
- UniGene ID
- Hs.531536
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_109773.1 |
|
3 |
372 |
|
GenBank mRNA |
BX338614.2 |
- |
3 |
367 |
|
- Gene Symbol
- HSD17B4
- Entrez Gene ID
- 3295
- Gene Name
- hydroxysteroid 17-beta dehydrogenase 4
- Gene Aliases
- DBP, MFE-2, MPF-2, PRLTS1, SDR8C1
- Chromosome Location
- Chr. 5: 119452443 - 119542335 on Build GRCh38
- UniGene ID
- Hs.406861
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL713635.1 |
- |
1 |
1367 |
|
- Gene Symbol
- IL1RAP
- Entrez Gene ID
- 3556
- Gene Name
- interleukin 1 receptor accessory protein
- Gene Aliases
- C3orf13, IL-1RAcP, IL1R3
- Chromosome Location
- Chr. 3: 190514051 - 190657197 on Build GRCh38
- UniGene ID
- Hs.478673
- Species
- Homo sapiens
- Gene Symbol
- KLHL36
- Entrez Gene ID
- 79786
- Gene Name
- kelch like family member 36
- Gene Aliases
- C16orf44
- Chromosome Location
- Chr. 16: 84648504 - 84667686 on Build GRCh38
- UniGene ID
- Hs.222731
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK122745.1 |
- |
1 |
1282 |
|
- Gene Symbol
- LINC00269
- Entrez Gene ID
- 100996279
- Gene Name
- long intergenic non-protein coding RNA 269
- Gene Aliases
- CXorf62, NCRNA00269
- Chromosome Location
- Chr. X: 69179557 - 69209924 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103715.1 |
|
5 |
663 |
|
GenBank mRNA |
AK090929.1 |
- |
5 |
663 |
|
- Gene Symbol
- LINC00887
- Entrez Gene ID
- 100131551
- Gene Name
- long intergenic non-protein coding RNA 887
- Gene Aliases
- Chromosome Location
- Chr. 3: 194301199 - 194312804 on Build GRCh38
- UniGene ID
- Hs.128066
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024480.1 |
|
3 |
1324 |
|
- Gene Symbol
- LOC100131792
- Entrez Gene ID
- 100131792
- Gene Name
- uncharacterized LOC100131792
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.638481
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK093561.1 |
- |
1 |
1219 |
|
- Gene Symbol
- LOC100507412
- Entrez Gene ID
- 100507412
- Gene Name
- uncharacterized LOC100507412
- Gene Aliases
- Chromosome Location
- Chr. Un|NT_167214: 97129 - 126696 on Build GRCh38
- UniGene ID
- Hs.426704
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038958.1 |
|
7 |
2159 |
|
GenBank mRNA |
AK057572.1 |
- |
7 |
2147 |
|
- Gene Symbol
- LOC102723753
- Entrez Gene ID
- 102723753
- Gene Name
- HECT and RLD domain containing E3 ubiquitin protein ligase 2 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 16|NT_187383: 1001630 - 1037618 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135178.1 |
|
3 |
372 |
|
GenBank mRNA |
AK300387.1 |
- |
3 |
372 |
|
DA859276.1 |
- |
3 |
372 |
|
- Gene Symbol
- NCOR1
- Entrez Gene ID
- 9611
- Gene Name
- nuclear receptor corepressor 1
- Gene Aliases
- N-CoR, N-CoR1, PPP1R109, TRAC1, hN-CoR
- Chromosome Location
- Chr. 17: 16030094 - 16216746 on Build GRCh38
- UniGene ID
- Hs.462323
- Species
- Homo sapiens
- Gene Symbol
- NEIL3
- Entrez Gene ID
- 55247
- Gene Name
- nei like DNA glycosylase 3
- Gene Aliases
- FGP2, FPG2, NEI3, ZGRF3, hFPG2, hNEI3
- Chromosome Location
- Chr. 4: 177309837 - 177362938 on Build GRCh38
- UniGene ID
- Hs.405467
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK307692.1 |
- |
4 |
1198 |
|
- Gene Symbol
- PDDC1
- Entrez Gene ID
- 347862
- Gene Name
- Parkinson disease 7 domain containing 1
- Gene Aliases
- Chromosome Location
- Chr. 11: 767095 - 777502 on Build GRCh38
- UniGene ID
- Hs.218362
- Species
- Homo sapiens
- Gene Symbol
- PPP2R3C
- Entrez Gene ID
- 55012
- Gene Name
- protein phosphatase 2 regulatory subunit B''gamma
- Gene Aliases
- C14orf10, G4-1, G5pr
- Chromosome Location
- Chr. 14: 35085467 - 35122543 on Build GRCh38
- UniGene ID
- Hs.530712
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC037797.1 |
- |
1 |
651 |
|
- Gene Symbol
- PPP4R3B
- Entrez Gene ID
- 57223
- Gene Name
- protein phosphatase 4 regulatory subunit 3B
- Gene Aliases
- FLFL2, PP4R3B, PSY2, SMEK2, smk1
- Chromosome Location
- Chr. 2: 55547292 - 55617757 on Build GRCh38
- UniGene ID
- Hs.516182
- Species
- Homo sapiens
- Gene Symbol
- PRKAR2A-AS1
- Entrez Gene ID
- 100506637
- Gene Name
- PRKAR2A antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 48847937 - 48851982 on Build GRCh38
- UniGene ID
- Hs.634259
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_109996.1 |
|
2 |
1298 |
|
NR_109997.1 |
|
3 |
1219 |
|
GenBank mRNA |
AK127190.1 |
- |
2 |
1278 |
|
- Gene Symbol
- RNF144A
- Entrez Gene ID
- 9781
- Gene Name
- ring finger protein 144A
- Gene Aliases
- RNF144, UBCE7IP4
- Chromosome Location
- Chr. 2: 6917392 - 7076886 on Build GRCh38
- UniGene ID
- Hs.22146
- Species
- Homo sapiens
- Gene Symbol
- RPL15
- Entrez Gene ID
- 6138
- Gene Name
- ribosomal protein L15
- Gene Aliases
- DBA12, EC45, L15, RPL10, RPLY10, RPYL10
- Chromosome Location
- Chr. 3: 23916804 - 23923696 on Build GRCh38
- UniGene ID
- Hs.381219
- Species
- Homo sapiens
- Gene Symbol
- SCLT1
- Entrez Gene ID
- 132320
- Gene Name
- sodium channel and clathrin linker 1
- Gene Aliases
- CAP-1A, CAP1A
- Chromosome Location
- Chr. 4: 128875206 - 129093607 on Build GRCh38
- UniGene ID
- Hs.654690
- Species
- Homo sapiens
- Gene Symbol
- SLC12A6
- Entrez Gene ID
- 9990
- Gene Name
- solute carrier family 12 member 6
- Gene Aliases
- ACCPN, KCC3, KCC3A, KCC3B
- Chromosome Location
- Chr. 15: 34229996 - 34338064 on Build GRCh38
- UniGene ID
- Hs.510939
- Species
- Homo sapiens
- Gene Symbol
- SPCS3
- Entrez Gene ID
- 60559
- Gene Name
- signal peptidase complex subunit 3
- Gene Aliases
- PRO3567, SPC22/23, SPC3, YLR066W
- Chromosome Location
- Chr. 4: 176319939 - 176332245 on Build GRCh38
- UniGene ID
- Hs.42194
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021928.3 |
NP_068747.1
|
5 |
4123 |
|
- Gene Symbol
- SPICE1
- Entrez Gene ID
- 152185
- Gene Name
- spindle and centriole associated protein 1
- Gene Aliases
- CCDC52, SPICE
- Chromosome Location
- Chr. 3: 113442718 - 113515187 on Build GRCh38
- UniGene ID
- Hs.477144
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144718.3 |
NP_653319.1
|
18 |
3832 |
|
GenBank mRNA |
AL832148.1 |
- |
18 |
3799 |
|
- Gene Symbol
- SYNE3
- Entrez Gene ID
- 161176
- Gene Name
- spectrin repeat containing nuclear envelope family member 3
- Gene Aliases
- C14orf49, NET53, Nesp3
- Chromosome Location
- Chr. 14: 95416083 - 95516693 on Build GRCh38
- UniGene ID
- Hs.354645
- Species
- Homo sapiens
- Gene Symbol
- TAF9B
- Entrez Gene ID
- 51616
- Gene Name
- TATA-box binding protein associated factor 9b
- Gene Aliases
- DN-7, DN7, TAF9L, TAFII31L, TFIID-31
- Chromosome Location
- Chr. X: 78129748 - 78139682 on Build GRCh38
- UniGene ID
- Hs.592248
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK074449.1 |
- |
1 |
1917 |
|
- Gene Symbol
- WDR55
- Entrez Gene ID
- 54853
- Gene Name
- WD repeat domain 55
- Gene Aliases
- Chromosome Location
- Chr. 5: 140664799 - 140674119 on Build GRCh38
- UniGene ID
- Hs.286261
- Species
- Homo sapiens
- Gene Symbol
- ZNF383
- Entrez Gene ID
- 163087
- Gene Name
- zinc finger protein 383
- Gene Aliases
- HSD17
- Chromosome Location
- Chr. 19: 37217243 - 37248740 on Build GRCh38
- UniGene ID
- Hs.590975
- Species
- Homo sapiens
View Full Product Details