Gene Transcripts

Gene Symbol
Olfr255
Entrez Gene ID
258185
Gene Name
olfactory receptor 255
Gene Aliases
GA_x6K02T2N0WB-1967-2905, MOR135-21P
Chromosome Location
Chr. : - on Build
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005780.1 NP_001005780.1 2 656

View Full Product Details