Gene Transcripts

Gene Symbol
KCNMA1
Entrez Gene ID
3778
Gene Name
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
Chromosome Location
Chr. 10: 76869601 - 77637819 on Build GRCh38
UniGene ID
Hs.144795
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014797.2 NP_001014797.1 29 3736
NM_001271518.1 NP_001258447.1 28 3574
NM_001322829.1 NP_001309758.1 29 3730
NM_001322830.1 NP_001309759.1 31 3826
NM_001322832.1 NP_001309761.1 28 3721
NM_001322835.1 NP_001309764.1 30 3814
NM_001322838.1 NP_001309767.1 30 3768
XM_005269776.3 XP_005269833.1 30 3820
XM_005269778.1 XP_005269835.1 30 3896
XM_005269781.2 XP_005269838.1 29 3819
XM_005269787.3 XP_005269844.1 29 3739
XM_005269789.2 XP_005269846.1 30 3738
XM_005269792.2 XP_005269849.1 29 3732
XM_005269796.2 XP_005269853.1 28 3645
XM_011539773.1 XP_011538075.1 31 4031
XM_011539775.1 XP_011538077.1 30 4019
XM_011539776.1 XP_011538078.1 30 4016
XM_011539780.1 XP_011538082.1 30 3899
XM_011539781.2 XP_011538083.1 30 3857
XM_011539785.1 XP_011538087.1 30 3800
XM_017016207.1 XP_016871696.1 32 4112
XM_017016208.1 XP_016871697.1 32 4109
XM_017016210.1 XP_016871699.1 31 4028
XM_017016211.1 XP_016871700.1 31 3941
XM_017016212.1 XP_016871701.1 31 3938
XM_017016213.1 XP_016871702.1 30 3929
XM_017016216.1 XP_016871705.1 30 3854
XM_017016217.1 XP_016871706.1 29 3794
XM_017016223.1 XP_016871712.1 27 3106
GenBank mRNA AB209488.1 - 28 3143
AK310379.1 - 30 3768
BX648925.1 - 25 2886

View Full Product Details