Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080437.1 | NP_001073906.1 | 32 | 5356 | |
XM_011510931.2 | XP_011509233.1 | 33 | 5670 | ||
XM_011510932.2 | XP_011509234.1 | 32 | 5573 | ||
GenBank mRNA | AF439717.1 | - | 26 | 4199 | |
AK074062.1 | - | 31 | 4964 | ||
AK074075.1 | - | 12 | 2683 |