Gene Transcripts

Gene Symbol
Endog
Entrez Gene ID
362100
Gene Name
endonuclease G
Gene Aliases
Chromosome Location
Chr. 3: 8741833 - 8744414 on Build Rnor_6.0
UniGene ID
Rn.3807
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034938.1 NP_001030110.1 2 599
GenBank mRNA AB221075.1 - 2 599
FQ232745.1 - 2 649

View Full Product Details