Gene Transcripts

Gene Symbol
CTTNBP2
Entrez Gene ID
83992
Gene Name
cortactin binding protein 2
Gene Aliases
C7orf8, CORTBP2, Orf4
Chromosome Location
Chr. 7: 117710648 - 117873507 on Build GRCh38
UniGene ID
Hs.592285
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033427.2 NP_219499.1 3 297
XM_005250634.4 XP_005250691.1 3 432
XM_005250635.3 XP_005250692.1 3 229
XM_011516615.2 XP_011514917.1 3 2600
XM_011516616.2 XP_011514918.1 3 229
XM_011516618.2 XP_011514920.1 3 229
XM_017012706.1 XP_016868195.1 3 229
XM_017012707.1 XP_016868196.1 3 229
GenBank mRNA AB051545.1 - 3 203
AF377960.1 - 3 297
AK091548.1 - 3 231
AK308081.1 - 3 229
BC106000.1 - 3 264 30528261

View Full Product Details