Gene Transcripts

Gene Symbol
CYB561
Entrez Gene ID
1534
Gene Name
cytochrome b561
Gene Aliases
CYB561A1, FRRS2
Chromosome Location
Chr. 17: 63432304 - 63446361 on Build GRCh38
UniGene ID
Hs.355264
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017916.1 NP_001017916.1 6 2204
NM_001017917.1 NP_001017917.1 6 2037
NM_001915.3 NP_001906.3 6 2034
XM_005257091.1 XP_005257148.1 6 1979
XM_017024260.1 XP_016879749.1 6 2076
XM_017024261.1 XP_016879750.1 5 1943
XM_017024262.1 XP_016879751.1 6 2269
GenBank mRNA AK095244.1 - 6 2251
BC002976.1 - 6 1991 3535771
BC091485.1 - 6 2005 6188259

Gene Symbol
PUM2
Entrez Gene ID
23369
Gene Name
pumilio RNA binding family member 2
Gene Aliases
PUMH2, PUML2
Chromosome Location
Chr. 2: 20248692 - 20352423 on Build GRCh38
UniGene ID
Hs.467824
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011532724.2 XP_011531026.1 14 11087

View Full Product Details