Gene Transcripts

Gene Symbol
Unc13d
Entrez Gene ID
192177
Gene Name
unc-13 homolog D (C. elegans)
Gene Aliases
Munc13-4, Unc13h4
Chromosome Location
Chr. 10: 104613907 - 104628664 on Build Rnor_6.0
UniGene ID
Rn.198919
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138844.1 NP_620199.1 4, 5 504
XM_006247702.2 XP_006247764.1 4, 5 689
GenBank mRNA AF159356.1 - 4, 5 504

View Full Product Details