Gene Transcripts

Gene Symbol
Kcnip4
Entrez Gene ID
80334
Gene Name
Kv channel interacting protein 4
Gene Aliases
AV032399, Calp, Calp250, KChIP4, KChIP4a
Chromosome Location
Chr. 5: 48389503 - 49525122 on Build GRCm38
UniGene ID
Mm.160172
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199242.1 NP_001186171.1 4 538
NM_001199243.1 NP_001186172.1 3 436
NM_001199244.1 NP_001186173.1 3 579
NM_001199245.1 NP_001186174.1 3 432
NM_030265.3 NP_084541.3 3 364
GenBank mRNA AF305071.1 - 3 335
AF453243.1 - 3 272
AK039048.1 - 3 365
AK049662.1 - 3 316
AK133416.1 - 3 284
AK148685.1 - 3 581
AK165773.1 - 3 306
AY647240.1 - 3 315
BC051130.1 - 4 538 6532082
DQ148501.1 - 4 538
DQ148502.1 - 2 296
DQ148503.1 - 3 316
DQ148504.1 - 3 432

View Full Product Details