Gene Transcripts

Gene Symbol
Zfand2a
Entrez Gene ID
100494
Gene Name
zinc finger, AN1-type domain 2A
Gene Aliases
AA407930, AU016206, Airap
Chromosome Location
Chr. 5: 139471216 - 139484491 on Build GRCm38
UniGene ID
Mm.24521
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159908.1 NP_001153380.1 4, 5 546
NM_133349.3 NP_579927.1 4, 5 557
GenBank mRNA AF224494.1 - 4, 5 443
AK146082.1 - 4, 5 495
AK171245.1 - 4, 5 497
AK198290.1 - 2, 3 174
BC058780.1 - 4, 5 483 6312202

View Full Product Details