Gene Transcripts
- Gene Symbol
- Tecpr2
- Entrez Gene ID
- 104859
- Gene Name
- tectonin beta-propeller repeat containing 2
- Gene Aliases
- 4930573I19Rik, AI604920, mKIAA0297
- Chromosome Location
- Chr. 12: 110889259 - 110972394 on Build GRCm38
- UniGene ID
- Mm.35583
- Species
- Mus musculus
- Gene Symbol
- Atf1
- Entrez Gene ID
- 11908
- Gene Name
- activating transcription factor 1
- Gene Aliases
- Chromosome Location
- Chr. 15: 100227859 - 100261248 on Build GRCm38
- UniGene ID
- Mm.676
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK148500.1 |
- |
|
1139 |
|
- Gene Symbol
- Cenpw
- Entrez Gene ID
- 66311
- Gene Name
- centromere protein W
- Gene Aliases
- 2610036L11Rik, Cug2
- Chromosome Location
- Chr. 10: 30196009 - 30200600 on Build GRCm38
- UniGene ID
- Mm.251779
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC050071.1 |
- |
|
3113 |
|
- Gene Symbol
- Csn3
- Entrez Gene ID
- 12994
- Gene Name
- casein kappa
- Gene Aliases
- AW208918, CSN10, Csnk
- Chromosome Location
- Chr. 5: 87925575 - 87932545 on Build GRCm38
- UniGene ID
- Mm.296303
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC004601.1 |
- |
3 |
976 |
|
BC005653.1 |
- |
2 |
1627 |
|
- Gene Symbol
- Ddx11
- Entrez Gene ID
- 320209
- Gene Name
- DEAD/H (Asp-Glu-Ala-Asp/His) box helicase 11
- Gene Aliases
- 4732462I11Rik, CHL1, CHLR1, KRG2, essa15a
- Chromosome Location
- Chr. 17: 66123501 - 66152174 on Build GRCm38
- UniGene ID
- Mm.259605
- Species
- Mus musculus
- Gene Symbol
- LOC105245566
- Entrez Gene ID
- 105245566
- Gene Name
- uncharacterized LOC105245566
- Gene Aliases
- Chromosome Location
- Chr. 13: 81782594 - 81783769 on Build GRCm38
- UniGene ID
- Mm.401912
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK144545.1 |
- |
|
1417 |
|
AK161157.1 |
- |
|
1397 |
|
- Gene Symbol
- Man2b1
- Entrez Gene ID
- 17159
- Gene Name
- mannosidase 2, alpha B1
- Gene Aliases
- AW107687, LAMAN
- Chromosome Location
- Chr. 8: 85082729 - 85098739 on Build GRCm38
- UniGene ID
- Mm.4219
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK054315.1 |
- |
4 |
3367 |
|
- Gene Symbol
- Ocstamp
- Entrez Gene ID
- 74614
- Gene Name
- osteoclast stimulatory transmembrane protein
- Gene Aliases
- 4833422F24Rik, OC-STAMP
- Chromosome Location
- Chr. 2: 165395450 - 165400394 on Build GRCm38
- UniGene ID
- Mm.72179
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK032330.1 |
- |
|
1535 |
|
- Gene Symbol
- Zfp874b
- Entrez Gene ID
- 408067
- Gene Name
- zinc finger protein 874b
- Gene Aliases
- 9630025I21Rik, Rslcan16
- Chromosome Location
- Chr. 13: 67471513 - 67486236 on Build GRCm38
- UniGene ID
- Mm.315879
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK034723.1 |
- |
|
2050 |
|
Chr. - - [-] |
View Full Product Details