Gene Transcripts
- Gene Symbol
- CPVL
- Entrez Gene ID
- 54504
- Gene Name
- carboxypeptidase, vitellogenic like
- Gene Aliases
- HVLP
- Chromosome Location
- Chr. 7: 28995630 - 29146537 on Build GRCh38
- UniGene ID
- Hs.233389
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019029.2 |
NP_061902.2
|
6, 7 |
743 |
|
NM_031311.3 |
NP_112601.3
|
6, 7 |
655 |
|
XM_011515437.1 |
XP_011513739.1
|
6, 7 |
822 |
|
XM_017012366.1 |
XP_016867855.1
|
6, 7 |
655 |
|
GenBank mRNA |
AF106704.2 |
- |
6, 7 |
575 |
|
AF217508.1 |
- |
5, 6 |
542 |
|
AF282617.1 |
- |
6, 7 |
605 |
|
AK075433.1 |
- |
6, 7 |
656 |
|
AK124472.1 |
- |
10, 11 |
1183 |
|
AK225545.1 |
- |
6, 7 |
606 |
|
AY358549.1 |
- |
6, 7 |
632 |
|
BC016838.2 |
- |
6, 7 |
703 |
3888647 |
View Full Product Details