Gene Transcripts

Gene Symbol
FAM153A
Entrez Gene ID
285596
Gene Name
family with sequence similarity 153 member A
Gene Aliases
NY-REN-7
Chromosome Location
Chr. 5: 177694247 - 177783518 on Build GRCh38
UniGene ID
Hs.368516
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173663.3 NP_775934.3 6 277
XM_005265889.3 XP_005265946.1 7 465
XM_006714848.3 XP_006714911.1 7 437
XM_006714849.3 XP_006714912.1 6 262
XM_011534520.2 XP_011532822.1 7 465
XM_011534521.2 XP_011532823.1 8 502
XM_011534522.2 XP_011532824.1 8 586
XM_011534523.2 XP_011532825.1 7 527
XM_017009352.1 XP_016864841.1 7 465
XM_017009353.1 XP_016864842.1 7 465
XM_017009354.1 XP_016864843.1 7 465
XM_017009355.1 XP_016864844.1 7 465
XM_017009356.1 XP_016864845.1 7 465
XM_017009357.1 XP_016864846.1 7 465
XM_017009358.1 XP_016864847.1 7 465
XM_017009359.1 XP_016864848.1 7 465
XM_017009360.1 XP_016864849.1 7 465
XM_017009361.1 XP_016864850.1 7 465
XM_017009362.1 XP_016864851.1 6 309
XM_017009363.1 XP_016864852.1 8 386
XM_017009364.1 XP_016864853.1 8 543
XM_017009365.1 XP_016864854.1 8 386
XM_017009366.1 XP_016864855.1 8 555
XM_017009367.1 XP_016864856.1 7 433
XM_017009368.1 XP_016864857.1 7 511
XM_017009369.1 XP_016864858.1 7 450
XM_017009370.1 XP_016864859.1 7 450
XM_017009371.1 XP_016864860.1 7 450
GenBank mRNA AB018295.1 - 6 238
AF155097.1 - 6 449
AK289518.1 - 8 586
DB043399.1 - 4 188
DB046513.1 - 6 345

Gene Symbol
FAM153B
Entrez Gene ID
202134
Gene Name
family with sequence similarity 153 member B
Gene Aliases
Chromosome Location
Chr. 5: 176063709 - 176114798 on Build GRCh38
UniGene ID
Hs.368516
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001265615.1 NP_001252544.1 6 277
GenBank mRNA BC028606.1 - 7 518

View Full Product Details