Gene Transcripts
- Gene Symbol
- Hipk2
- Entrez Gene ID
- 15258
- Gene Name
- homeodomain interacting protein kinase 2
- Gene Aliases
- 1110014O20Rik, B230339E18Rik, Stank
- Chromosome Location
- Chr. 6: 38687512 - 38876190 on Build GRCm38
- UniGene ID
- Mm.23790
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001136065.2 |
NP_001129537.1
|
6, 7 |
1922 |
|
NM_001294143.1 |
NP_001281072.1
|
6, 7 |
1998 |
|
NM_001294144.1 |
NP_001281073.1
|
6, 7 |
1998 |
|
NM_010433.2 |
NP_034563.2
|
6, 7 |
1998 |
|
GenBank mRNA |
AF077659.1 |
- |
6, 7 |
1761 |
|
AF170301.1 |
- |
6, 7 |
1748 |
|
AF170302.1 |
- |
6, 7 |
1748 |
|
AF208292.1 |
- |
6, 7 |
1835 |
|
AF273680.1 |
- |
6, 7 |
1719 |
|
AF333791.1 |
- |
6, 7 |
1625 |
|
AF333792.1 |
- |
6, 7 |
1625 |
|
AK019821.1 |
- |
2, 3 |
214 |
|
BC031904.1 |
- |
6, 7 |
1731 |
5368577 |
View Full Product Details