Gene Transcripts

Gene Symbol
Stk25
Entrez Gene ID
59041
Gene Name
serine/threonine kinase 25 (yeast)
Gene Aliases
1500019J11Rik, AU018434, C86992, Ste20-like, Ysk1
Chromosome Location
Chr. 1: 93620751 - 93654216 on Build GRCm38
UniGene ID
Mm.28761
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021537.3 NP_067512.3 7 885
GenBank mRNA AF004934.1 - 6 723
AK148041.1 - 7 859
AK152917.1 - 7 828
BC029875.1 - 6 1603
BC052913.1 - 7 820 30057101
BC071218.1 - 7 880 4012695

View Full Product Details