Gene Transcripts

Gene Symbol
Chrm5
Entrez Gene ID
53949
Gene Name
cholinergic receptor, muscarinic 5
Gene Aliases
Chromosome Location
Chr. 3: 103966451 - 104018815 on Build Rnor_6.0
UniGene ID
Rn.44293
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017362.4 NP_059058.1 1661
GenBank mRNA M22926.1 - 1661

View Full Product Details