Gene Transcripts

Gene Symbol
Mmp8
Entrez Gene ID
63849
Gene Name
matrix metallopeptidase 8
Gene Aliases
Chromosome Location
Chr. 8: 5768808 - 5777741 on Build Rnor_6.0
UniGene ID
Rn.44474
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022221.1 NP_071557.1 3 440
XM_006242496.2 XP_006242558.1 3 521
GenBank mRNA AJ007288.1 - 3 440

View Full Product Details